missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™

Function tested and optimized for LC/MS applications

Brand:  JT Baker 9830-02

Additional Details : CAS Number : 67-56-1 Weight : 1.60000kg Transport : UN number : 1230 Chem class : 3 Pack group : II

Packaging Narrow Mouth Amber Glass Bottle
Quantity 1L

 View more versions of this product

Product Code. 15548664

  • 79.25 € / 6 L

Local Stock 4

Please to purchase this item. Need a web account? Register with us today!


Description

Description

    For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use
Specifications

Chemical Identifiers

67-56-1
32.042
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol

Specifications

Methanol
67-56-1
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.042
CHEBI:17790
BAKER ANALYZED™ LC-MS Reagent
1L = 0.80kg
1L
Filtered through a 0.2 micron filter
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Narrow Mouth Amber Glass Bottle
12.00°C
Videos
Documents