missing translation for 'onlineSavingsMsg'
Learn More

Methanol, HPLC for Gradient Analysis, Fisher Chemical™

Catalog Number 10000280
78.25 EUR valid until 2024-12-31
Use promo code "21964" to get your promotional price.


64.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.


74.87 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.


78.25 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle
67-56-1
CH4O
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO

Methanol, HPLC for Gradient Analysis

Specification
Molecular Formula: CH4O
Water Soluble: Yes
Molecular Weight: 32.04
Melting Point: -98°C
Health Hazards: Causes moderate eye irritation, may cause skin irritation, may cause irritation of the digestive tract
For Use With [Application]: HPLC for gradient analysis
Flash Point: 12°C
CAS No.: 67-56-1
Boiling Point: 64.7°C
Hazard Symbol: F, T
Synonyms: Carbinol, Methyl Alcohol, Methyl Hydroxide, Monohydroxymethane, Wood Alcohol, Wood Naptha, Wood Spirits, Columbian Spirits
TRUSTED_SUSTAINABILITY
Melting Point -98°C
Color Colorless
Boiling Point 64.7°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55cp at 20°C
Molecular Weight (g/mol) 32.04
Formula Weight 32.04g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'