missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, for HPLC-MS, Fisher Chemical™
35.63 EUR valid until 2024-12-31
Use promo code "21964" to get your promotional price.
15.71 EUR valid until 2024-03-30
Use promo code "21663" to get your promotional price.
Use promo code "21964" to get your promotional price.
15.71 EUR valid until 2024-03-30
Use promo code "21663" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
Methanol, for HPLC-MS
Specifications
Melting Point | -98°C |
Color | Colorless |
Formula Weight | 32.04g/mol |
Boiling Point | 64.7°C |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 128 hPa at 20°C |
Viscosity | 0.55cp at 20°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'