missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Ultra Gradient Grade, BAKER ANALYZED HPLC, J.T. Baker™

Catalog Number 10423861
Click to view available options
Quantity:
5 L
Packaging:
EcoTainer Metal Safety Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Get rapid, reproducible performance and separation results. Methanol, Ultra Gradient Grade, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use
Melting Point -98°C
Absorbance Max. 0.20 at 225nm, Max. 0.02 at 254nm, Max. 0.01 at 280nm, Max. 0.01 at 350nm
CAS 67-56-1
Molecular Formula CH4O
Linear Formula CH3OH
MDL Number MFCD00004595
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Solubility Information Fully miscible in water
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Percent Purity ≥99.8%
Grade Ultra Gradient Grade
Density 0.79g/cm3
Boiling Point 64.7°C
Quantity 5 L
Explosion Limit 5.5 - 44 Vol %
Flash Point 11°C
Ignition Point 455°C
Odor Alcohol-like
Packaging EcoTainer Metal Safety Can
Physical Form Liquid
Residue after Evaporation Max. 2 ppm
Titratable Acid Max. 0.0003meq/g
Titratable Base Max. 0.0001meq/g
Water Max. 0.02%
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'