missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate-d3 Heptaglutamate Trifluoroacetate, TRC

Product Code. 30477809
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477809 10 mg 10mg
30478521 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30477809 Supplier Toronto Research Chemicals Supplier No. TRCM26069210MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C50H61D3N14O23 xC2HF3O2
Molecular Weight (g/mol) 1232.142
SMILES NC1=NC(N)=NC2=C1N=C(CN(C([2H])([2H])[2H])C3=CC=C(C(N[C@H](C(O)=O)CCC(O)=O)=O)C=C3)C=N2.O=C(O)C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H22N8O5.C2HF3O2/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30;3-2(4,5)1(6)7/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27);(H,6,7)/t13-;/m0./s1/i1D3;
Chemical Name or Material Methotrexate-d3 Heptaglutamate Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.