missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dimethoxybenzoate, 98%, Thermo Scientific™

Catalog Number 10021314
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2150-41-6
C10H12O4
196.20
MFCD00070649
IHIJFZWLGPEYPJ-UHFFFAOYSA-N
137451
methyl 2,4-dimethoxybenzoate
COC(=O)C1=CC=C(OC)C=C1OC
This item is not returnable. View return policy
2150-41-6
C10H12O4
196.20
MFCD00070649
IHIJFZWLGPEYPJ-UHFFFAOYSA-N
137451
methyl 2,4-dimethoxybenzoate
COC(=O)C1=CC=C(OC)C=C1OC
Density 1.1940g/mL
Boiling Point 294°C to 296°C
Flash Point >110°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH30)2C6H3CO2CH3
Quantity 100 g
Specific Gravity 1.194
Synonym 2,4-dimethoxybenzoic acid methyl ester, benzoic acid, 2,4-dimethoxy-, methyl ester, acmc-1ccii, methyl2,4-dimethoxybenzoate, methyl 2,4-dimethoxybenzoate, 2,4-dimethoxy-benzoic acid methyl ester, benzoic acid,2,4-dimethoxy-,methyl ester, benzoic acid,2,4-dimethoxy-, methyl ester, 2,4-dihydroxybenzoic acid, dimethyl ether, methyl ester
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material Methyl 2, 4-dimethoxybenzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'