missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate, 97%, Thermo Scientific™

Catalog Number p-7077323
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
175137-27-6
C7H4ClF3N2O2
240.57
MFCD00068146
VLUBJYUOPNGBOQ-UHFFFAOYSA-N
methyl 2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, methyl 2-chloro-4-trifluoromethyl-5-pyrimidinecarboxylate, methylchlorotrifluoromethylpyrimidinecarboxylate, methyl-2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, acmc-20aifi, pubchem10174, 2-chloro-4-trifluoromethyl-5-carbomethoxypyrimidine, methyl 2-chloro-4-trifluoromethyl pyrimidine-5-ca, 2-chloro-5-methoxycarbonyl-4-trifluoromethylpyrimidine, 2-chloro-5-methoxycarbonyl-4-trifluoromethyl pyrimidine
3696469
methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
COC(=O)C1=CN=C(Cl)N=C1C(F)(F)F
This item is not returnable. View return policy
175137-27-6
C7H4ClF3N2O2
240.57
MFCD00068146
VLUBJYUOPNGBOQ-UHFFFAOYSA-N
methyl 2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, methyl 2-chloro-4-trifluoromethyl-5-pyrimidinecarboxylate, methylchlorotrifluoromethylpyrimidinecarboxylate, methyl-2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, acmc-20aifi, pubchem10174, 2-chloro-4-trifluoromethyl-5-carbomethoxypyrimidine, methyl 2-chloro-4-trifluoromethyl pyrimidine-5-ca, 2-chloro-5-methoxycarbonyl-4-trifluoromethylpyrimidine, 2-chloro-5-methoxycarbonyl-4-trifluoromethyl pyrimidine
3696469
methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
COC(=O)C1=CN=C(Cl)N=C1C(F)(F)F
Boiling Point 56.0°C (0.2 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 240.57
Percent Purity 97%
Chemical Name or Material Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
compliance-icons
Product Identifier
  • Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'