missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dichlorobenzoate, Tech., Maybridge™

Brand:  Maybridge KM02512DA

Code : 31

Additional Details : CAS Number : 2905-67-1 Weight : 0.05100kg

Quantity 1g

 View more versions of this product

Product Code. 10025002

  • 9.85 € / 1g

Estimated Shipment
01-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2905-67-1
205.034
3,5-dichlorobenzoic acid methyl ester, benzoic acid, 3,5-dichloro-, methyl ester, unii-4gim06ee3p, 4gim06ee3p, methyl-3,5-dichlorobenzoate, maybridge1_004559, acmc-1cd9x, dsstox_cid_22146, dsstox_rid_79937, dsstox_gsid_42146
methyl 3,5-dichlorobenzoate
C8H6Cl2O2
BTEVDFJXGLQUDS-UHFFFAOYSA-N
76192
COC(=O)C1=CC(=CC(=C1)Cl)Cl

Specifications

Methyl 3, 5-dichlorobenzoate
2905-67-1
C8H6Cl2O2
BTEVDFJXGLQUDS-UHFFFAOYSA-N
methyl 3,5-dichlorobenzoate
76192
Tech.
1g
90%
3,5-dichlorobenzoic acid methyl ester, benzoic acid, 3,5-dichloro-, methyl ester, unii-4gim06ee3p, 4gim06ee3p, methyl-3,5-dichlorobenzoate, maybridge1_004559, acmc-1cd9x, dsstox_cid_22146, dsstox_rid_79937, dsstox_gsid_42146
COC(=O)C1=CC(=CC(=C1)Cl)Cl
205.034
205.04
Technical
Videos
SDS Documents