missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dichlorobenzoate, Tech., Thermo Scientific™

Catalog Number p-4391402
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
2905-67-1
C8H6Cl2O2
205.03
MFCD00032710
BTEVDFJXGLQUDS-UHFFFAOYSA-N
3,5-dichlorobenzoic acid methyl ester, benzoic acid, 3,5-dichloro-, methyl ester, unii-4gim06ee3p, 4gim06ee3p, methyl-3,5-dichlorobenzoate, maybridge1_004559, acmc-1cd9x, dsstox_cid_22146, dsstox_rid_79937, dsstox_gsid_42146
76192
methyl 3,5-dichlorobenzoate
COC(=O)C1=CC(Cl)=CC(Cl)=C1
This item is not returnable. View return policy
2905-67-1
C8H6Cl2O2
205.03
MFCD00032710
BTEVDFJXGLQUDS-UHFFFAOYSA-N
3,5-dichlorobenzoic acid methyl ester, benzoic acid, 3,5-dichloro-, methyl ester, unii-4gim06ee3p, 4gim06ee3p, methyl-3,5-dichlorobenzoate, maybridge1_004559, acmc-1cd9x, dsstox_cid_22146, dsstox_rid_79937, dsstox_gsid_42146
76192
methyl 3,5-dichlorobenzoate
COC(=O)C1=CC(Cl)=CC(Cl)=C1
Assay Percent Range 90%
Quantity 1 g
Formula Weight 205.04
Percent Purity Tech.
Grade Technical
Chemical Name or Material Methyl 3, 5-dichlorobenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'