missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hexenoate, 95+%, predominantly trans, ACROS Organics™

Brand:  Acros Organics 393550250

Code : 29

Additional Details : CAS Number : 2396-78-3 Weight : 0.07500kg Transport : UN number : 3272 Chem class : 3 Pack group : III


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10014973

  • 26.02 € / 25g

Estimated Shipment
02-02-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2396-78-3
128.17
XEAIHUDTEINXFG-SNAWJCMRSA-N
5362782
CCC=CCC(=O)OC
C7H12O2
MFCD00672771
3-hexenoic acid, methyl ester, methyl 3-hexenoate, methyl trans-3-hexenoate, methyl e-hex-3-enoate, honeyflor, 3-hexenoic acid, methyl ester, e, unii-7ip0t834pb, methyl hex-3-enoate, methyl e-3-hexenoate, methyl trans 3 hexenoate
methyl (E)-hex-3-enoate

Specifications

Methyl 3-hexenoate
48°C
Glass bottle
0.915
Colorless to Yellow
2396-78-3
C7H12O2
MFCD00672771
Solubility in water: practically insoluble
CCC=CCC(=O)OC
128.17
128.17
95+%
0.9150g/mL
Authentic
1.4250 to 1.4270 (20°C, 589nm)
169°C
25g
95% min. (GC)
CH3CH2CHCHCH2COOCH3
3-hexenoic acid, methyl ester, methyl 3-hexenoate, methyl trans-3-hexenoate, methyl e-hex-3-enoate, honeyflor, 3-hexenoic acid, methyl ester, e, unii-7ip0t834pb, methyl hex-3-enoate, methyl e-3-hexenoate, methyl trans 3 hexenoate
XEAIHUDTEINXFG-SNAWJCMRSA-N
methyl (E)-hex-3-enoate
5362782
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl 3-hexenoate, predominantly trans

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents