Learn More
Methyl 5-iodosalicylate 99%, ACROS Organics™
Brand: Acros Organics 381770100
Code : 29
Additional Details : CAS Number : 4068-75-1 Weight : 0.06000kg
Packaging | Glass Bottle |
---|---|
Quantity | 10g |
Chemical Identifiers
4068-75-1 | |
278.045 | |
NRSWJTRJHPRZMH-UHFFFAOYSA-N | |
248910 | |
COC(=O)C1=C(C=CC(=C1)I)O |
C8H7IO3 | |
MFCD00016462 | |
methyl 5-iodosalicylate, 5-iodosalicylic acid methyl ester, 2-hydroxy-5-iodobenzoic acid methyl ester, benzoic acid, 2-hydroxy-5-iodo-, methyl ester, methyl 2-hydroxy-5-iodobenzenecarboxylate, salicylic acid, 5-iodo-, methyl ester, methyl5-iodosalicylate, methyl-5-iodosalicylate, acmc-209jg0, methyl2-hydroxy-5-iodobenzoate | |
methyl 2-hydroxy-5-iodobenzoate |
Specifications
Methyl 5-iodosalicylate | |
Glass Bottle | |
73°C to 78°C | |
4068-75-1 | |
C8H7IO3 | |
MFCD00016462 | |
NRSWJTRJHPRZMH-UHFFFAOYSA-N | |
methyl 2-hydroxy-5-iodobenzoate | |
248910 | |
Crystalline Powder |
Authentic | |
White | |
10g | |
99% | |
IC6H3(OH)COOCH3 | |
methyl 5-iodosalicylate, 5-iodosalicylic acid methyl ester, 2-hydroxy-5-iodobenzoic acid methyl ester, benzoic acid, 2-hydroxy-5-iodo-, methyl ester, methyl 2-hydroxy-5-iodobenzenecarboxylate, salicylic acid, 5-iodo-, methyl ester, methyl5-iodosalicylate, methyl-5-iodosalicylate, acmc-209jg0, methyl2-hydroxy-5-iodobenzoate | |
COC(=O)C1=C(C=CC(=C1)I)O | |
278.045 | |
278.04 | |
≥98.5% |
Safety and Handling
- Methyl 5-iodosalicylate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.