missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl butyrate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
Methyl butyrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl butyrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
Methyl butyrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
Specifications
Melting Point | -85°C to -84°C |
Boiling Point | 100°C to 102°C |
Flash Point | 11°C (51°F) |
Quantity | 100 mL |
UN Number | UN1237 |
Beilstein | 1740743 |
Merck Index | 14,6035 |
Formula Weight | 102.14 |
Percent Purity | ≥98% |
Chemical Name or Material | Methyl butyrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'