missing translation for 'onlineSavingsMsg'
Learn More

Methyl cellulose, viscosity 1600 cPs

Product Code. 11304828
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 11304828

Brand: Thermo Scientific Alfa Aesar 043146.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

Thickener and emulsifier in various food and cosmetic products, and also as a treatment of constipation. Methyl cellulose, as a gel, has the unique property of setting when hot and melting when cold. Methyl cellulose is used as a variable viscosity personal lubricant. Methyl cellulose is used in the manufacture of capsules in nutritional supplements. Methyl cellulose finds a major application as a performance additive in construction materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Odor Odorless
Quantity 500 g
Merck Index 14,6040
Solubility Information Readily Soluble in a water below 50°C and wide variety of solvents.
Physical Form Powder
Chemical Name or Material Methyl cellulose, Viscosity: 1600cPs

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.