missing translation for 'onlineSavingsMsg'
Learn More

Methyl formate, 97%, pure, Thermo Scientific Chemicals

Catalog Number p-163359
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
107-31-3
C2H4O2
60.05
MFCD00003291
TZIHFWKZFHZASV-UHFFFAOYSA-N
formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
7865
CHEBI:77699
methyl formate
COC=O
107-31-3
C2H4O2
60.05
MFCD00003291
TZIHFWKZFHZASV-UHFFFAOYSA-N
formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
7865
CHEBI:77699
methyl formate
COC=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -100.0°C
Color Colorless
Density 0.9680g/mL
Boiling Point 31.0°C to 33.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HCO2CH3
Refractive Index 1.3420 to 1.3440
Quantity 1 L
Beilstein 02,18
Fieser 05,446
Merck Index 15,6148
Specific Gravity 0.968
Solubility Information Solubility in water: 300g/L (20°C). Other solubilities: soluble in almost all conventional organic solvent,miscible with alcohol,soluble in ether
Viscosity 0.35 mPa.s (20°C)
Formula Weight 60.05
Percent Purity 97%
Grade Pure
Physical Form Liquid
Chemical Name or Material Methyl formate, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'