missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl tert-butylacetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
Description
Methyl tert-butylacetate is an oxygenated solvent used alone or in blends for coatings, industrial cleaning and surface treatment. It is used as an intermediate to manufacture pharmaceuticals, synthetic flavorings and other organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl tert-butylacetate is an oxygenated solvent used alone or in blends for coatings, industrial cleaning and surface treatment. It is used as an intermediate to manufacture pharmaceuticals, synthetic flavorings and other organic
Methyl tert-butylacetate is an oxygenated solvent used alone or in blends for coatings, industrial cleaning and surface treatment. It is used as an intermediate to manufacture pharmaceuticals, synthetic flavorings and other organic
Specifications
CAS | 10250-48-3 |
Boiling Point | 126°C to 128°C |
Flash Point | 24°C (75°F) |
Molecular Formula | C7H14O2 |
Refractive Index | 1.3995 |
MDL Number | MFCD00043646 |
Quantity | 10 g |
UN Number | UN3272 |
Beilstein | 1747895 |
Synonym | methyl tert-butylacetate, methyl 3,3-dimethylbutyrate, butanoic acid, 3,3-dimethyl-, methyl ester, butyric acid, 3,3-dimethyl-, methyl ester, methyl t-butylacetate, acmc-1c3ge, methyl-tert-butyl acetate, 3,3-dimethylbutyric acid methyl ester, 3,3-dimethyl-butyric acid methyl ester |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'