missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet 2B, Thermo Scientific Chemicals

Catalog Number p-3776823
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
8004-87-3
C24H28ClN3
MFCD00012568
Basic Violet 1, C.I. 42535
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
8004-87-3
C24H28ClN3
MFCD00012568
Basic Violet 1, C.I. 42535

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 137.0°C
Absorption Ratio 1.06 to 1.19
Dye Content 75% min.
Beilstein 13, 755
Solubility Information Solubility in water: soluble. Other solubilities: soluble in conc. sulfuric acid and nitric acid, soluble in chloroform, partially soluble in alcohol and glycerol
Infrared Spectrum Authentic
Lambda Max 583 to 587nm
Formula Weight 393.95
Grade Pure
Packaging Glass bottle
Color Green
Quantity 25 g
Physical Form Crystalline Powder With Metal Luster
Chemical Name or Material Methyl Violet 2B, Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'