missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-formylbenzoate, 98%, Thermo Scientific Chemicals

Catalog Number p-2691745
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
52178-50-4
C9H8O3
164.2
MFCD00189379
UVSBCUAQEZINCQ-UHFFFAOYSA-N
3-carbomethoxybenzaldehyde, methyl m-formylbenzoate, benzoic acid, 3-formyl-, methyl ester, 3-formylbenzoic acid methyl ester, methyl-3-formylbenzoate, pubchem12631, m-carbomethoxybenzaldehyde, methyl 3-formyl-benzoate, methyl 3-methanoylbenzoate, acmc-1am8j
639145
methyl 3-formylbenzoate
COC(=O)C1=CC=CC(=C1)C=O
This item is not returnable. View return policy
52178-50-4
C9H8O3
164.2
MFCD00189379
UVSBCUAQEZINCQ-UHFFFAOYSA-N
3-carbomethoxybenzaldehyde, methyl m-formylbenzoate, benzoic acid, 3-formyl-, methyl ester, 3-formylbenzoic acid methyl ester, methyl-3-formylbenzoate, pubchem12631, m-carbomethoxybenzaldehyde, methyl 3-formyl-benzoate, methyl 3-methanoylbenzoate, acmc-1am8j
639145
methyl 3-formylbenzoate
COC(=O)C1=CC=CC(=C1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 48.0°C to 55.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material Methyl 3-formylbenzoate, 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'