missing translation for 'onlineSavingsMsg'
Learn More

Methyl pivaloylacetate, 98%, ACROS Organics™

Brand:  Acros Organics 414190010

Code : 29

Additional Details : CAS Number : 55107-14-7 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11945111

  • 16.37 € / 1g

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

55107-14-7
158.2
methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2
methyl 4,4-dimethyl-3-oxopentanoate
C8H14O3
XTXCFTMJPRXBBC-UHFFFAOYSA-N
99597
CC(C)(C)C(=O)CC(=O)OC

Specifications

Methyl pivaloylacetate
78°C
Glass Bottle
0.98
Colorless
55107-14-7
C8H14O3
methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2
CC(C)(C)C(=O)CC(=O)OC
158.2
158.2
98%
0.9800g/mL
Authentic
1.4300 to 1.4320
67.0°C to 70.0°C (13.0mmHg)
1g
97.5% min. (GC)
(CH3)3CCOCH2COOCH3
XTXCFTMJPRXBBC-UHFFFAOYSA-N
methyl 4,4-dimethyl-3-oxopentanoate
99597
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl pivaloylacetate, 98%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates