missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Methylthymol blue, tetrasodium salt, pure, water soluble
88.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
5g; Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Indicator
Specifications
CAS | 1945-77-3 |
Molecular Formula | C37H44N2Na4O13S |
MDL Number | MFCD00148905 |
Beilstein | 19, V,8, 619 |
Merck Index | 15, 6202 |
Synonym | 3,3'-Bis[N,N-di(carboxymethyl)aminomethyl]thymolsulfonephthalein, sodium salt, MTB |
InChI Key | LGVVZVZPOQJZKT-UHFFFAOYSA-N |
Infrared Spectrum | Authentic |
SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] |
IUPAC Name | 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'