missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium Bromide, 98%, ACROS Organics™

Brand:  Acros Organics 156950025

Code : 29

Additional Details : CAS Number : 1779-49-3 Weight : 2.55000kg


Packaging Plastic bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 11975801

  • 631.00 € / 2.50kg

Estimated Shipment
28-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

1779-49-3
357.22
LSEFCHWGJNHZNT-UHFFFAOYSA-M
74505
C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
C19H18BrP
MFCD00011804
methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide
methyl(triphenyl)phosphanium;bromide

Specifications

Methyltriphenylphosphonium bromide
Authentic
White
2.5kg
97.5% min. (Argentometry)
CH3P(C6H5)3Br
01,697
Solubility in water: 400g/L (25°C). Other solubilities: 950g/L methanol (20°C),soluble in aliphatic alcohols and chloroform,practically insoluble in arom. hydrocarbons and,acetone
C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
357.22
357.22
98%
>240°C
Plastic bottle
230.0°C to 234.0°C
1779-49-3
C19H18BrP
MFCD00011804
methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide
LSEFCHWGJNHZNT-UHFFFAOYSA-M
methyl(triphenyl)phosphanium;bromide
74505
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyltriphenylphosphonium bromide

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents