Learn More
Methylurea, 97%, ACROS Organics™
Brand: Acros Organics 127995000
Code : 29
Additional Details : CAS Number : 598-50-5 Weight : 0.55000kg
Packaging | Plastic bottle |
---|---|
Quantity | 500g |
Chemical Identifiers
598-50-5 | |
74.08 | |
XGEGHDBEHXKFPX-UHFFFAOYSA-N | |
11719 | |
methylurea |
C2H6N2O | |
MFCD00007950 | |
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea | |
CHEBI:44383 | |
CNC(=O)N |
Specifications
Methylurea | |
Plastic bottle | |
White | |
500g | |
598-50-5 | |
100.0 | |
C2H6N2O | |
MFCD00007950 | |
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea | |
XGEGHDBEHXKFPX-UHFFFAOYSA-N | |
methylurea | |
11719 | |
74.08 | |
97% |
Authentic | |
2% max. | |
98.0°C to 102.0°C | |
97% | |
96.0 | |
96% min. (on dry substance) (HPLC) | |
CH3NHCONH2 | |
04, 64 | |
Solubility in water: 1000g/L (20°C). Other solubilities: freely soluble in ethanol | |
CNC(=O)N | |
74.08 | |
CHEBI:44383 | |
Crystalline Solid |