Learn More
Emma Safety Footwear Metric XD Safety Shoes
Unique microfiber upper is fully washable. Features SmartecAir™ for dry, fresh feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors often seen in high-tech and cleanroom environments.
Brand:  Emma Safety Footwear 507560-42
Additional Details : Weight : 1.39500kg
 View more versions of this product
Product Code. 15988621
Quantity Price
1 80.20 € / 1 pair
10 73.15 € / 1 pair
Estimated Shipment: 12-11-2021
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
Metric XD
Microfiber
42