missing translation for 'onlineSavingsMsg'
Learn More

Metro™ High-Density qwikTRAK™ Track Sets

Create tracks up to 21 ft.

Brand:  Metro™ BTS9NA

Additional Details : Weight : 4.72000kg

 View more versions of this product

Product Code. 15986891

  • 959.90 € / Each

Estimated Shipment: 08-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q and MetroMax i shelving systems
  • Sets include tracks and hardware needed to assemble system up to 21 ft. (640cm)
  • 17-823-715 (BTS2.5NA) includes a joining kit to connect the track to other track sets

Specifications

Specifications

274.32 cm
Tracks and hardware needed to assemble high-density qwikTRAK™ system up to 21 ft.
5 kg
Track Set
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q™ and MetroMax i™ shelving system
13 lb.
108 in.
Videos
SDS
Documents
Certificates