missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax i™ Lab Worktable Accessory, Drawer
The Metro MetroMax i™ Lab Worktable Drawer comes in a choice of sizes to provide extra room and functionality in your storage setup.
229.00 € - 399.00 €
Product Code | Brand | Depth (English) | Depth (Metric) | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (English) | Depth (Metric) | Product Type | Price | Quantity & Availability | |||
15917061
|
Metro™
LTD3L |
3 in. | 7.6 cm | Locking Drawer |
329.00 €
Each |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||
15907061
|
Metro™
LTD3 |
3 in. | 7.6 cm | Drawer |
229.00 €
Each |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||
15927061
|
Metro™
LTD6 |
6 in. | 15.2 cm | Drawer |
299.00 €
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15937061
|
Metro™
LTD6L |
6 in. | 15.2 cm | Locking Drawer |
399.00 €
Each |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||
Description
- Drawers fit 30, 36, 48 and 60 in. (76, 91.4, 121.9, and 152.4cm) table lengths
- Locking feature available
Specifications
For 30, 36, 48 and 60 in. lab worktables |