Learn More
Metro™ MetroMax i™ Lab Worktable, Stainless Top and Backsplash with Solid i Shelf
The Metro MetroMax i™ Lab Worktable advanced polymer construction provides excellent corrosion-resistance performance for the most corrosive lab environments.
Brand:  Metro™ LTM30XS3
Additional Details : Weight : 28.30400kg
 View more versions of this product
Product Code. 15987061
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Table features smooth surfaces that are easy to clean
  • Mobile units can be easily moved to clean behind table area
  • Available top materials include Type 304 stainless and TRESPA phenolic resin (black or gray)
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
29.75 in.
29.75 x 30 x 39.5 in. (74.9 x 76.2 x 100.3 cm)
100.3 cm
Backsplash with solid Metro™ MetroMax i™ shelf, adaptor insert
273 kg
74.9 cm
Polymer
Mobile Table
Ideal for lab
39.5 in.
30 in.
76.2 cm
600 lb.