missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax i™ Top-Track Mobile Unit Kits
High-density, easy access storage
461.00 € - 805.00 €
Specifications
For Use With (Equipment) | Metro™ MetroMax i™ and Q™ Top-Track™ system |
---|---|
Product Type | Mobile Unit Kit |
Product Code | Brand | Includes | Width (English) Shelf | Width (Metric) Shelf | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Width (English) Shelf | Width (Metric) Shelf | Price | Quantity & Availability | |||
15957221
|
Metro™
MXTTM18C |
Posts, Chrome plated rigid casters, caster channel, bumpers and roller assemblies | 18 in. | 45.7 cm |
461.00 €
Each |
Estimated Shipment: 18-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15967221
|
Metro™
MXTTM18S |
Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies | 18 in. | 45.7 cm |
780.00 €
Each |
Estimated Shipment: 18-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15977221
|
Metro™
MXTTM24C |
Posts, Chrome plated rigid casters, caster channel, bumpers and roller assemblies | 24 in. | 61 cm |
485.00 €
Each |
Estimated Shipment: 18-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15987221
|
Metro™
MXTTM24S |
Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies | 24 in. | 61 cm |
805.00 €
Each |
Estimated Shipment: 18-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
- Mobile units move along anchored track allowing easy access to all shelves
Order Info
- Order one per mobile unit
- Shelves sold separately
Specifications
Metro™ MetroMax i™ and Q™ Top-Track™ system | |
Mobile Unit Kit |