missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax iQ™ Accessory, Keyboard Shelf
For use with MetroMax iQ shelves
Brand: Metro™ LTKB
Additional Details : Weight : 2.90400kg
Description
- Epoxy-coated shelf provides high level of corrosion resistance
- Mounts to 18 in. or 24 in. wide shelves
- Minimum shelf length is 30 in.
Shelf load should not exceed 10 lbs. (45.8kg).
Specifications
48 in. (121.9mm) and 60 in. (152.4mm) wide MetroMax i shelves | |
Keyboard Shelf |