missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax™ iQ Polymer Shelving Accessory, Color Shelf Marker
Use to assign color coding to individual shelf levels, shelving units, carts, or strorage areas
6.75 €
Specifications
For Use With (Equipment) | Metro™ MetroMax i™ and Q™ shelving |
---|---|
Length (English) | 6 in. |
Length (Metric) | 15.2 cm |
Description
- Attaches easity to shelf for content identification
- 6 in. length
Specifications
Metro™ MetroMax i™ and Q™ shelving | |
6 in. | |
15.2 cm |