missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MW Series Utility Cart with Wire Shelves

Made of Super Erecta™ shelves with plastic split sleeves, two one-piece handles and casters with donut bumpers, Metro MW Series Utility Cart with Wire Shelves make a smart addition to any workplace.

Brand:  Metro™ MW708

Additional Details : Weight : 26.51200kg

 View more versions of this product

Product Code. 15927181

  • 1465.00 € / Each

Estimated Shipment: 14-03-2023
to see stock.
Add to basket

Description

Description

Cart

  • Load capacity: 375 lb. (170.1kg)
  • Fully interchangeable Super Erecta shelves
  • Center shelf can be positioned at any height in 1 in. (2.5cm) increments
  • One-piece stainless-steel or chrome handles (depends on model) add rigidity and durability
  • 18 in. (46cm) carts have 4 in. dia. (10cm) casters; 21 in.(53cm) and 24 in. (61cm) wide carts have 5 in. dia. (13cm) casters
  • Three tiers
  • 375 lbs (170kg) weight capacity per cart
Wire Shelves
  • Chrome-plated or stainless-steel shelves available with trademarked truss design for added strength
  • Corners slide easily over special tapered snap-together plastic sleeves to fasten them securely in place

Order Info

Shipped knocked down. Assembles/disassembles quickly and easily without tools.

Specifications

Specifications

Utility Cart
3-tier Model
Transporting materials that require an expanse of flat, solid surface
39 in.
Super Erecta™ 3 Stainless Steel wire shelves with Plastic split sleeves, 2 one-piece Stainless Steel Handle, 5 in. dia. caster, 3.5 in. donut bumpers
91 cm
170 kg
Stainless Steel
53 cm
NSF
36 x 21 x 39 in. (91 x 53 x 99 cm)
Stainless Steel
99 cm
36 in.
375 lb.
3
21 in.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers