Learn More
Metro™ Stainless Lab Worktable, Black Phenolic Top and Solid HD Shelf
Safeguard your workers in the laboratory. Metro Stainless Lab Worktable with a solid HD shelf and a stainless top/backsplash provides corrosion-proof performance and stainless steel construction throughout.
Brand: Metro™ LTS60PB
Additional Details : Weight : 51.16800kg
Description
- Easily cleaned smooth surfaces
- Mobile units can be easily moved to clean behind table area
- Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
59.75 x 30 x 36 in. (151.7 x 76.2 x 91.4 cm) | |
36 in. | |
Solid HD shelf | |
151.7 cm | |
30 in. | |
600 lb. | |
273 kg |
Ideal for lab | |
91.4 cm | |
59.75 in. | |
Stainless Steel | |
76.2 cm | |
Stationary Table |