Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Configure your lab storage as needed. Metro Super Erecta Wire Shelves are easily adjustable and the Metroseal 3 green epoxy with Microban finish means they are easy to keep clean. Shelves are for use with Metro Super Erecta shelving systems.
Marca:  Metro™ 1824NK3
67.98 EUR valid until 2024-11-01
Use promo code "21705" to get your promotional price.
 Ver más versiones de este producto
Código de producto. 15916591
  • 94.15 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore más ofertas especiales
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
Descripción
Descripción
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Corrosion resistant finish for wet or high humidity environments
  • Antimicrobial is built into finish to keep product clean
Especificaciones
Especificaciones
18 x 24 in.
Metroseal 3 with Microban
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales