Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Configure your lab storage as needed. Metro Super Erecta Wire Shelves are easily adjustable and the Metroseal 3 green epoxy with Microban finish means they are easy to keep clean. Shelves are for use with Metro Super Erecta shelving systems.
Brand:  Metro™ 3660NK3
200.76 EUR valid until 2024-11-01
Use promo code "21705" to get your promotional price.
 View more versions of this product
Product Code. 15946701
  • 278.00 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Corrosion resistant finish for wet or high humidity environments
  • Antimicrobial is built into finish to keep product clean
Specifications
Specifications
36 x 60 in.
Metroseal 3 with Microban