Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Configure your lab storage as needed. Metro Super Erecta Wire Shelves are easily adjustable and the Metroseal 3 green epoxy with Microban finish means they are easy to keep clean. Shelves are for use with Metro Super Erecta shelving systems.
Brand:  Metro™ 1448NK3
Additional Details : Weight : 3.44800kg
 View more versions of this product
Product Code. 15996571
  • 104.00 € / Each
Estimated Shipment: 03-09-2024
to see stock.
Add to basket
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Corrosion resistant finish for wet or high humidity environments
  • Antimicrobial is built into finish to keep product clean
Specifications
Specifications
14 x 48 in.
Metroseal 3 with Microban