missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Wire Shelving Track Sets

For use with Metro Super Erecta™, Super Erecta Pro™ and Super Adjustable™ Top Track™ shelving

Brand:  Metro™ TTS10NA

Additional Details : Weight : 26.85600kg

 View more versions of this product

Product Code. 15987301

  • 900.00 € / Each
Estimated Shipment: 05-09-2024
to see stock.
Add to basket

Description

Description

  • Track sets include necessary sections of track for assembling track runs up to 21 ft. (640cm)
  • Coated with Microban™ antimicrobial protection

Order Info

19-138-228 (TTS2.5NA) includes a joining kit to connect the track to other track sets.

Specifications

Specifications

Necessary sections of track for assembling track runs of 1830 to 6405mm
120 in.
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ top track shelving
304.8 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers