Learn More
Metro™ MetroMax Shelf Posts
Compatible with MetroMax i™ and MetroMax Q™ shelves
38.60 € - 59.50 €
Specifications
For Use With (Equipment) | Metro™ MetroMax i™ and Q™ shelving |
---|---|
Material | Polymer |
Product Code | Brand | Height (English) | Height (Metric) | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (English) | Height (Metric) | Product Type | Prix | Quantité et disponibilité | |||
15957211
|
Metro™
MX74P |
74 in. | 188 cm | Stationary Post with Leveling Foot |
51.70 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
15967211
|
Metro™
MX74UP |
74 in. | 188 cm | Mobile Post for Stem Caster |
54.90 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||
15917211
|
Metro™
MX33UP |
33 in. | 83.82 cm | Mobile Post for Stem Caster |
38.60 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
15907211
|
Metro™
MX33P |
33 in. | 83.82 cm | Stationary Post with Leveling Foot |
38.60 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
15947211
|
Metro™
MX63P |
63 in. | 160 cm | Stationary Post with Leveling Foot |
45.15 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
15937211
|
Metro™
MX54UP |
54 in. | 137.1 cm | Mobile Post for Stem Caster |
42.70 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||
15977211
|
Metro™
MX86P |
86 in. | 218.4 cm | Stationary Post with Leveling Foot |
59.50 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
Description
Grooves located every 1 in. for locking shelf wedges in place. Post height stamped to face of post for easy shelf positioning.
- Corrosion-proof polymer posts with built-in Microban™ antimicrobial product protection
- Stationary posts include an adjustable leveling foot to compensate for uneven floors
- Leveling foot can be adjusted 1 in. (25mm)
- Mobile posts accept optional Metro Stem Casters, available in a variety of materials to meet your specific needs
Specifications
Metro™ MetroMax i™ and Q™ shelving | |
Polymer |