missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ ISA Solutions

Solutions for perfectION™ combined ion selective electrodes

Brand:  Mettler Toledo™ 51344760

Code : 59

Additional Details : Weight : 0.64000kg

 View more versions of this product

Product Code. 11947511

  • 95.40 € / 475mL


Estimated Shipment
02-07-2021
Add to basket

Description

Description

Solutions (ionic-strength adjustment buffers)

  • Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
  • ISA for calcium electrode, 475mL
  • ISA for potassium electrode, 475mL
  • ISA for nitrate electrode, 475mL
  • Nitrate ISS (for suppressing interference), 475mL
  • Fluoride TISAB II with CDTA, 3790mL
  • Fluoride TISAB III with CDTA (concentrate), 475mL

Specifications

Specifications

perfectION ISA solid state ISE
475 mL
ISA Solution
Ion Electrolyte B (Chloride, copper, iodide, silver)
475 mL
Videos
SDS
Documents