missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters

Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.

Brand:  METTLER TOLEDO™ 51302621

Code : 59

Additional Details : Weight : 2.28000kg

 View more versions of this product

Product Code. 10042274

  • 2325.00 € / Each
Currently Unavailable: We have no information when this product will be available
View alternative products

Description

Description

  • Dust proof
  • Water proof
  • IP67 Rated
  • pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
  • Compatible with a wide range of IP67-rated electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Data storage available; capacity dependent on model
  • Export data to a printer or Excel file for Seven2Go Pro only
  • EasyDirect software available for data transfer (model dependent) for Seven2Go Pro only
  • Rubber holster available for single-handed measurements
  • uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters
Specifications

Specifications

IR to printer or PC via RS232 or USB
±0.1mV
±0.5%
5 Points, 7 pre-defined buffer group
LCD Graphic, Backlit
0.01 μS/cm to 1000 mS/cm
-2 to 19.999
0 to 80 ppt
0.1/0.01/0.001
InLab 738-ISM conductivity/temperature electrode; InLab Expert Go-ISM pH/temperature electrode
Dual Channel; pH/ORP/ion, conductivity
Electrode Kit
0.1mV
±0.2°C
±0.5%
±0.002
4 x AA
ISO 9001, ISO 14001
IP 67
-1999 to 1999 mV
0 to 100megohm/cm
0.01 mg/L to 300 g/L
Determination of pH, mV, rel. mV or ion concentrations and conductivity, TDS (total dissolved solids), salinity or specific resistance
pH: BNC/RCA, conductor: LTW 7-pin
NTC
0.01/1
0.01/1
0.1°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers