Learn More
Merck MF-Millipore™ Membrane Filter, 0.45 μm, Gridded
Mixed cellulose esters (MCE) membrane, hydrophilic
Brand: Merck HAWG03700
Code : 71
Additional Details : Weight : 0.00500kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
Not Autoclavable | |
White | |
Cellulose Acetate, Cellulose Nitrate | |
Non-sterile | |
Water, air monitoring, analytical and research applications | |
Hydrophilic | |
100 | |
Filter Membranes | |
37 mm |
≥1.8 bar, air with water (at 23°C) | |
Gravimetric Analysis Membrane | |
0.45 μm | |
60 mL/min x cm2 | |
4 L/min x cm2 | |
100Pack | |
150 μm | |
Hydrophilic | |
37 mm |
Safety and Handling
- Monitor Refills .45 MCE WH GRD 37mm 100pk
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use powder to extinguish.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose