missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Micro Filter Funnels
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
40.12 € - 41.02 €
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
11586764
|
Lenz Laborglasintrumente
07190110 |
1 |
40.12 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11516774
|
Lenz Laborglasintrumente
07190410 |
4 |
41.02 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11576764
|
Lenz Laborglasintrumente
07190010 |
0 |
40.12 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
- Stem dia.: 6 mm
- Material: borosilicate glass 3.3
Specifications
2 mL | |
Borosilicate Glass 3.3 |