Learn More
Emma Safety Footwear Micro Safety Shoes
Unique microfiber upper is fully washable. SmartecAir™ provides fresh dry feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors used in high-tech and cleanroom environments.
Brand:  Emma Safety Footwear 500540-43
Additional Details : Weight : 1.41400kg
 View more versions of this product
Product Code. 15988451
Quantity Price
1 79.25 € / 1 pair
10 72.30 € / 1 pair
Estimated Shipment: 22-11-2021
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
White
Hydro-Tec™ Sanitized Silver
43
Elastic
Micro
Microfiber
Steel