Learn More
Ansell Edmont™ Microchem™ 3000 Coveralls with Hood and Integrated Socks
MICROCHEM™ 3000 Coveralls with Hood and Integrated Socks is one of the lightest and most comfortable chemical protective garments on the market today. This durable multi-layer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
Brand:  Ansell Edmont™ YE30W-00122-03
Additional Details : Weight : 0.39000kg
 View more versions of this product
Product Code. 15546594
  • 655.00 € / Pack of 20
Estimated Shipment
05-02-2020
Add to basket
Description
Description
  • Coverall with 2-piece hood
  • Integrated socks with boot overflap
  • Double zip closure
  • Double cuffs
  • Elasticated hood, waist, double cuffs
  • Excellent liquid penetration resistance and barrier to fine particulates (>0.01 microns)
  • Moisture vapour permeable (“breathable”) to help reduce the risk of heat stress
  • Silicone free for paint spraying applications
  • Low linting to reduces the risk of fibre contamination in some critical areas
  • Optimised body fit for wearer comfort and safety
  • Anti-static according to EN 1149-5
Specifications
Specifications
Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5
Welded
Medium
Includes Hood, Integrated socks with boot overflap
Protective coverall
Elastic, Attached socks, Boot overflaps
Hood, integrated socks with boot overflap
0
Double, Elastic
Yellow
2-piece, elasticated
Elasticated
Double-zip
Hooded
Multi-layer barrier laminate