Learn More
Lenz Laborglasinstrumente™ Microfilter Funnels
Created from borosilicate glass. Lenz Laborglasinstrumente™ Microfilter Funnels are ideal for concentrating organisms in fluid samples.
Brand:  Lenz Laborglasinstrumente™ 07190310
Additional Details : Weight : 0.10000kg
Product Code. 15111985
  • 40.55 € / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Made of borosilicate glass
  • Filter out microorganisms
Specifications
Specifications
Borosilicate Glass
3
2 mL
10 mm
1