missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Microflex™ 93-852 Series Single Use Nitrile Gloves

Provides contrasting backdrop for identifying lighter colored material. Ansell Edmont™ Microflex™ 93-852 Series Single Use Nitrile Gloves has distinct color to mask stains for special applications.

Brand:  Ansell Edmont™ 93852080

Additional Details : Weight : 0.57904kg

 View more versions of this product

Product Code. 15549471

Quantity Price
1 13.00 € / Pack of 100
10 11.48 € / Pack of 100

UK Stock 7
Add to basket


Description

Description

  • Single use nitrile gloves
  • Offers higher tensile strength compared to both ASTM D6319 and EN 455 standards, allowing the hands to move freely and comfortably
  • Fully textured design
  • Non-foaming formula enable wearers to have firm wet grip when handling objects
  • Contains no natural rubber latex
  • Powder-free
  • Helps protect wearers from Type I skin allergies, skin irritation and dryness

Applications:

  • Analytical testing/Measurements
  • Biotechnologies
  • Filtration processes
  • Food Processing
  • Laboratory analysis
  • Laboratory testing
  • Light assembly tasks
  • Light assembly of oil-coated pieces
  • Light duty maintenance and clean up
  • Mechanical jobs
  • Paint and spray shops
  • Pharmaceutical manufacturing
  • Protection from blood and other bodily fluids
Specifications

Specifications

Grip design: Textured surface
Yes
100 gloves per dispenser, 10 dispensers per shipper carton, 1000 gloves per shipper carton
7.5 - 8
Nitrile gloves
Powder-free
245mm
Beaded
Nitrile
93-852
0.12mm
Single use gloves
Black
EN 420:2003 + A1:2009 and EN 374:2003
Videos
Documents