missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) MicroFunnel™ Filters Funnel with Petri Dish Lid Kit and GN-6 Membrane

Ready-to-use, sterile, individually bagged with a lid kit to culture-in-place, reduce chances of cross contamination.

Brand:  Cytiva (formerly Pall Lab) 4810

Additional Details : Weight : 1.61500kg

Product Code. 17154411

  • 360.00 € / Pack of 50
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Easy squeeze removal of funnel to access membrane
  • No twisting means less RSI and membrane tearing
  • For research use only
Specifications

Specifications

Sterile
0.45 μm
Gridded
Metricel Mixed Cellulose Ester
50/Pk.
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers