missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) MicroFunnel™ Filters Funnel with Polycarbonate Membrane

Funnel ideal for concentrating organisms in fluid samples.

Brand:  Cytiva (formerly Pall Lab) FMFNL3020

Additional Details : Weight : 1.71000kg

 View more versions of this product

Product Code. 17114731

  • 156.00 € / Pack of 20
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

  • Flat, smooth surface allows for easy elution of organisms or particulate
  • Ready-to-use, disposable design eliminates sample cross contamination
Specifications

Specifications

Sterile
300 mL
Polycarbonate Membrane
0.4 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers