Learn More
Ansell Edmont™ Microgard™ 1500 Plus Blue Coveralls with Hood
Provides a good barrier against particulates such as asbestos, brick dust and cement dust. Ansell Edmont™ Microgard™ 1500 Plus Blue Coveralls with Hood filter 99.9% of particles >3μ.
Brand:  Ansell Edmont™ BL15S-00111-07
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 15546424
  • 212.00 € / Pack of 30
Estimated Shipment
23-12-2019
Add to basket
Description
Description
  • Coverall with 3-piece hood, stitched seams, elasticated hood, waist, wrists and ankles
  • 2-way front zipper with re-sealable stormflap
  • Anti-static treated SMS fabric
  • Breathable to help reduce the risk of heat stress 
  • Silicone-free which is critical in spray painting applications
  • Optimized body fit for wearer comfort and safety
Specifications
Specifications
EN ISO 13982-1, EN13034, EN1073-2, EN1149-5
Stitched
3X-Large
Includes Hood, re-sealable storm flap
Protective coverall
Elastic
Hood, re-sealable storm flap
0
Elastic
Blue
3-piece, elasticated
Elasticated
2-way front zipper
Hooded
Anti-static treated SMS fabric