missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microglass Fiber Filter Discs
Brand: Fisherbrand™
Code : 47
Additional Details : Weight : 0.04000kg
Description
- High loading capacity
- Made from 100% borosilicate glass
- Compatible with many solvents and reagents
- Contain no binder
- Not affected by humidity
- Suitable for use at and up to 500°C
Specifications
75 g/m2 | |
Disc | |
0.7 μm | |
Air pollution control, separation of soot before gas analyzers, support weighing in the balance analyzes and dryers | |
Non-hygroscopic | |
0.017 in. | |
Filter |
Microfilter | |
Borosilicate Glass | |
310 sec. | |
263 | |
500°C | |
0.45 mm | |
90 mm |