missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microglass Fiber Filter Discs
Made with 100% borosilicate micro-glass fibers. Fisherbrand™ Microglass Fiber Filter Discs are non-hygroscopic and biologically inert. They show chemical resistance to most solvents and reagents (except hydrofluoric acid).
47.35 € - 120.00 €
Specifications
Basis Weight | 75 g/m2 |
---|---|
Filter Type | Microfilter |
Format | Disc |
Pore Size | 0.7 μm |
Filtration Time | 310 sec. |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11724626
|
Fisherbrand™
|
109 mm |
85.95 €
Pack of 50 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
11708563
|
Fisherbrand™
|
70 mm |
73.65 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11714626
|
Fisherbrand™
|
90 mm |
66.15 €
Pack of 50 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11778553
|
Fisherbrand™
|
55 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11784093
|
Fisherbrand™
|
124 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11735265
|
Fisherbrand™
|
37 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Contains no binder
- Suitable for use at and up to 500°C
- Not affected by humidity
- Suitable for micro-filtration due to fine fibers
- High-loading capacity
- High chemical purity
- made with demineralized water
- High permeability to air
Applications:
- Filtration of industrial water, fresh water algae, drinking water, Proteins, samples for HPLC, solvents
- Sediments content in petroleum products
- Ligand binding test
- Air pollution monitoring
- Membranes pre-filters
- Radio-immuno tests
- Suspended solids in water, smokes
- Liquid scintillation counting
- Cells collection
- Grease content in dairy products
Specifications
75 g/m2 | |
Disc | |
310 sec. | |
Non-hygroscopic | |
0.45 mm |
Microfilter | |
0.7 μm | |
263 | |
500°C | |
Filter |