missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microglass Fiber Filters, Grade 259
Made with 100% borosilicate micro-glass fibers. Fisherbrand™ Microglass Fiber Filters, Grade 259 are non-hygroscopic and biologically inert. They show chemical resistance to most solvents and reagents (except hydrofluoric acid).
16.20 € - 94.45 €
Specifications
Basis Weight | 52 g/m2 |
---|---|
Format | Disc |
Material | Borosilicate Glass |
Pore Size | 1.6 μm |
Filtration Time | 60 sec. |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11475238
|
Fisherbrand™
|
109 mm |
60.05 €
Pack of 100 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11774073
|
Fisherbrand™
|
124 mm |
60.05 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11784073
|
Fisherbrand™
|
149 mm |
94.45 €
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
11714073
|
Fisherbrand™
|
25 mm |
16.20 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11724073
|
Fisherbrand™
|
45.7 mm |
50.95 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11798762
|
Fisherbrand™
|
55 mm |
32.35 €
Pack of 100 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11455238
|
Fisherbrand™
|
70 mm |
31.30 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11465238
|
Fisherbrand™
|
90 mm |
39.70 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11765245
|
Fisherbrand™
|
99 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11745245
|
Fisherbrand™
|
37 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11758553
|
Fisherbrand™
|
42.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11748553
|
Fisherbrand™
|
38 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Contains no binder
- Suitable for use at and up to 500°C
- Not affected by humidity
- Suitable for micro-filtration due to fine fibers
- High-loading capacity
- High chemical purity
- made with demineralized water
- High permeability to air
Applications:
- Filtration of industrial water, fresh water algae, drinking water, Proteins, samples for HPLC, solvents
- Sediments content in petroleum products
- Ligand binding test
- Air pollution monitoring
- Membranes pre-filters
- Radio-immuno tests
- Suspended solids in water, smokes
- Liquid scintillation counting
- Cells collection
- Grease content in dairy products
Specifications
52 g/m2 | |
Borosilicate Glass | |
60 sec. | |
0.26 mm |
Disc | |
1.6 μm | |
259 |