missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Microscope Slides with Ground Edges

Examine objects with steadiness. Perfect for sturdy examination of objects under a microscope.

Brand:  Hirschmann™ 8210101

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10009680

  • 84.20 € / Pack of 1000


Estimated Shipment
13-07-2020
Add to basket

Description

Description

  • Ground Edges 90°C ISO 8037/l
  • With LOT number on packaging
  • CE acc. to IVD 98/79 EC
Specifications

Specifications

0.95 to 1.05 mm
ISO 8037/1, CE, IVD 98/79 EC
8210101
Extra White
Ground edges 90°
76 x 26 mm
Videos
Documents